2,3,5,6-Tetrafluorophenol, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
196789-25G 196789-5G
SIAL196789-25GEA 153 EUR
SIAL196789-25G SIAL196789-5G
2,3,5,6-Tetrafluorophenol, Sigma-Aldrich®
2,3,5,6-Tetrafluorophenol
Formula: C₆H₂F₄O
Boiling Pt: 140 °C (1013 hPa)
Melting Pt: 30 °C
MDL Number: MFCD00002157
CAS Number: 769-39-1
EINECS: 212-209-6
UN: 3261
ADR: 8,II