Pentafluorophenol, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 2,3,4,5,6-Pentafluorophenol
M-Clarity™ quality level = MQ100
103799-100G 103799-10G 103799-5G
SIAL103799-100GEA 297 EUR
SIAL103799-100G SIAL103799-10G SIAL103799-5G
Pentafluorophenol, Sigma-Aldrich®
Pentafluorophenol
Formula: C₆HF₅O
Boiling Pt: 143 °C (1013 hPa)
Melting Pt: 34…36 °C
Density: 1.75 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002156
CAS Number: 771-61-9
EINECS: 212-235-8
UN: 3261
ADR: 8,II