3-Fluorobenzenesulphonyl chloride ≥98%
Supplier: Thermo Fisher Scientific
Danger
A13140.03 A13140.06
A13140.03EA 32.5 EUR
A13140.03 A13140.06
3-Fluorobenzenesulphonyl chloride ≥98%
3-Fluorobenzenesulphonyl chloride
Purity: 98% CAS No.: 701-27-9 Molecular Formula: C6H4ClFO2S Formula Weight: 194.61 Flash Point: 110°(230°F) Refractive Index: 1.5290 Melting Point: 7° Boiling Point: 231-232° Beilstein Reference: 2091631 UN No.: 3265 MDL No.: MFCD00042286
Formula: C₆H₄ClFO₂S
Boiling Pt: 231…232 °C
Melting Pt: 7 °C
Density: 1.463
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042286
CAS Number: 701-27-9
UN: 3265
ADR: 8,II