3-(Dimethylamino)phenol ≥97%
Supplier: Thermo Fisher Scientific
|
Warning
|
B23067.06
B23067.14
B23067.22
B23067.06EA
29.6
EUR
B23067.06
B23067.14
B23067.22
3-(Dimethylamino)phenol ≥97%
3-(Dimethylamino)phenol
Purity: 97+%
CAS No.: 99-07-0
Molecular Formula: C8H11NO
Formula Weight: 137.18
Flash Point: 148°(298°F)
Odour: Pungent
Boiling Point: 267°
Beilstein Reference: 774631
MDL No.: MFCD00002264
Formula:
C₈H₁₁NO Boiling Pt: 267 °C Melting Pt: 80…84 °C Flash Pt: 148 °C (298 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002264 CAS Number: 99-07-0 EINECS: 202-727-0 |