2-Ethoxyphenyl isocyanate

Supplier: Thermo Scientific

Synonyms: 1-Ethoxy-2-isocyanatobenzene

SB01858.1 SB01858.10 SB01858.50
MAYBSB01858.1EA 0 EUR
MAYBSB01858.1 MAYBSB01858.10 MAYBSB01858.50
2-Ethoxyphenyl isocyanate
2-Ethoxyphenyl isocyanate
Formula: C₉H₉NO₂
Boiling Pt: 106…110 °C (11 mmHg)
Density: 1.1 g/cm³ (20 °C)
MDL Number: MFCD00002007
CAS Number: 5395-71-1
UN: 3334
ADR: 9,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR