4-Nitrophenyl isothiocyanate

Supplier: Thermo Scientific
Warning

AC41607.1 AC41607.50
MAYBAC41607.1EA 0 EUR
MAYBAC41607.1 MAYBAC41607.50
4-Nitrophenyl isothiocyanate
4-Nitrophenyl isothiocyanate
Formula: C₇H₄N₂O₂S
Boiling Pt: 331 °C (1013 hPa)
Melting Pt: 108…110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007307
CAS Number: 2131-61-5
EINECS: 218-359-9
UN: 2923
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR