2-Nitroaniline, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: o-Nitroaniline
M-Clarity™ quality level = MQ200
N9780-100G N9780-500G N9780-5G
SIALN9780-100GEA 89.8 EUR
SIALN9780-100G SIALN9780-500G SIALN9780-5G
2-Nitroaniline, Sigma-Aldrich®
2-Nitroaniline
Formula: C₆H₆N₂O₂
MW: 138.13 g/mol
Boiling Pt: 284
Melting Pt: 68…71 °C
Density: 1.446 g/cm³ (20 °C)
Flash Pt: 167
Storage Temperature: Ambient
MDL Number: MFCD00007687
CAS Number: 88-74-4
EINECS: 201-855-4
UN: 1661
ADR: 6.1 II
Merck Index: 12,06680