2-Amino-4-nitrophenol, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
A70402-25G A70402-5G
SIALA70402-25GEA 49.1 EUR
SIALA70402-25G SIALA70402-5G
2-Amino-4-nitrophenol, Sigma-Aldrich®
2-Amino-4-nitrophenol
Formula: C₆H₆N₂O₃
Melting Pt: 176…177 °C
Density: 1.66 g/cm³ (20 °C)
MDL Number: MFCD00007695
CAS Number: 99-57-0
EINECS: 202-767-9