3-Aminophenol, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 3-Hydroxyaniline, m-Aminophenol
M-Clarity™ quality level = MQ200
100242-100G 100242-500G 100242-5G
SIAL100242-100GEA 48.5 EUR
SIAL100242-100G SIAL100242-500G SIAL100242-5G
3-Aminophenol, Sigma-Aldrich®
3-Aminophenol
Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 299
Melting Pt: 122
Density: 1.275 g/cm³ (20 °C)
Flash Pt: 92
MDL Number: MFCD00007786
CAS Number: 591-27-5
EINECS: 209-711-2
UN: 2512
ADR: 6.1 III
Merck Index: 13,00459