4-Aminothiophenol 96%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
4-Aminobenzenethiol, 4-Aminophenyl mercaptan, 4-Mercaptoaniline
104680050
104680250
ACRO104680050EA
52.9
EUR
ACRO104680050
ACRO104680250
4-Aminothiophenol 96%
4-Aminothiophenol
Formula:
C₆H₇NS Boiling Pt: 140…145 °C (16 mmHg) Melting Pt: 37…43 °C Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007880 CAS Number: 1193-02-8 EINECS: 214-763-4 UN: 3263 ADR: 8,III |
Specification Test Results
Appearance | Yellow Low melting crystalline mass |
Infrared spectrum | Conforms |
Melting point | 37 to 43 °C |
Titration Argentometric | ≥95 % |
Additional info | oxidized (disulphide) compound is steadily formed in air |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...