4-Aminothiophenol 97%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: 4-Aminobenzenethiol, 4-Aminophenyl mercaptan, 4-Mercaptoaniline
A14082.06 A14082.14 A14082.22
A14082.06EA 47.3 EUR
A14082.06 A14082.14 A14082.22
4-Aminothiophenol 97%
4-Aminothiophenol
Purity: 97% CAS No.: 1193-02-8 Molecular Formula: C6H7NS Formula Weight: 125.19 Flash Point: 110°(230°F) Odour: Stench Boiling Point: 156-157° Beilstein Reference: 906904 UN No.: 3263 MDL No.: MFCD00007880
Formula: C₆H₇NS
Boiling Pt: 156…157 °C
Melting Pt: 37…42 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007880
CAS Number: 1193-02-8
EINECS: 214-763-4
UN: 3263
ADR: 8,III