4-Nitro-o-phenylenediamine, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 4-Nitro-1,2-phenylenediamine, 4-Nitrobenzene-1,2-diamine
M-Clarity™ quality level = MQ100
108898-100G 108898-5G
SIAL108898-100GEA 88.1 EUR
SIAL108898-100G SIAL108898-5G
4-Nitro-o-phenylenediamine, Sigma-Aldrich®
4-Nitro-o-phenylenediamine
Formula: C₆H₇N₃O₂
MW: 153.14 g/mol
Melting Pt: 197…202 °C
MDL Number: MFCD00007724
CAS Number: 99-56-9
EINECS: 202-766-3
Merck Index: 12,06720