Diphenylamine, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: N-Phenylaniline, N-Phenylbenzeneamine
M-Clarity™ quality level = MQ200
242586-100G 242586-500G 242586-5G
SIAL242586-100GEA 76.5 EUR
SIAL242586-100G SIAL242586-500G SIAL242586-5G
Diphenylamine, Sigma-Aldrich®
Diphenylamine
Formula: (C₆H₅)₂NH
MW: 169.23 g/mol
Boiling Pt: 302
Melting Pt: 53…54 °C
Density: 1.165 g/cm³ (20 °C)
Flash Pt: 153 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003014
CAS Number: 122-39-4
EINECS: 204-539-4
UN: 3077
ADR: 9 III
Merck Index: 13,03349
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23b05bdc0d251be7093ced24f3f9389bc8e8d3c5&vid=f543cfadbe22d37b18bd37917bb8b5672a6d2df4&prid=web&clid=SAPDAM