1,2-Dinitrobenzene, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
126632-25G 126632-5G
SIAL126632-25GEA 257 EUR
SIAL126632-25G SIAL126632-5G
1,2-Dinitrobenzene, Sigma-Aldrich®
1,2-Dinitrobenzene
Formula: C₆H₄N₂O₄
Boiling Pt: 297 °C (1013 hPa)
Melting Pt: 112…115 °C
Density: 1.31 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007093
CAS Number: 528-29-0
EINECS: 208-431-8
UN: 3443
ADR: 6.1,II