4-Nitrobenzenesulphonyl chloride
Supplier: Thermo Scientific
Danger
Synonyms: 4-Nitrobenzene-1-sulphonyl chloride
SB01738.1 SB01738.50
MAYBSB01738.1EA 0 EUR
MAYBSB01738.1 MAYBSB01738.50
4-Nitrobenzenesulphonyl chloride
4-Nitrobenzenesulphonyl chloride
Formula: C₆H₄ClNO₄S
Boiling Pt: 350 °C (1013 hPa)
Melting Pt: 79 °C
Density: 1.69 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007445
CAS Number: 98-74-8
EINECS: 202-697-9
UN: 3261
ADR: 8,II