4-Methoxythiophenol, Sigma-Aldrich®
Supplier: Merck
Synonyms: p-Methoxythiophenol, 4-Mercaptoanisol, 1-Methoxy-4-mercaptobenzene, 4-Methoxybenzenethiol, 4-Methoxybenzene-1-thiol, 4-Methoxyphenyl mercaptan
M-Clarity™ quality level = MQ200
109525-25G 109525-5G
SIAL109525-25GEA 270 EUR
SIAL109525-25G SIAL109525-5G
4-Methoxythiophenol, Sigma-Aldrich®
4-Methoxythiophenol
Formula: C₇H₈OS
Boiling Pt: 228 °C (1013 hPa)
Melting Pt: 42 °C
Density: 1.1397 g/cm³ (20 °C)
MDL Number: MFCD00004849
CAS Number: 696-63-9
EINECS: 211-799-2
UN: 2810
ADR: 6.1,III