Veratrole, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
140155-500G 140155-1KG 140155-100G
SIAL140155-500GEA 128 EUR
SIAL140155-500G SIAL140155-1KG SIAL140155-100G
Veratrole, Sigma-Aldrich®
Veratrole
Formula: C₈H₁₀O₂
Boiling Pt: 207 °C (1013 hPa)
Melting Pt: 19…21 °C
Density: 1.085 g/cm³ (20 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00008357
CAS Number: 91-16-7
EINECS: 202-045-3
Merck Index: 12,10089