Veratrole, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ100
W379905-1KG
W379905-5KG
W379905-SAMPLE
SIALW379905-1KGEA
116
EUR
SIALW379905-1KG
SIALW379905-5KG
SIALW379905-SAMPLE
Veratrole, Sigma-Aldrich®
Veratrole
Formula:
C₈H₁₀O₂ Boiling Pt: 207 °C (1013 hPa) Melting Pt: 19…21 °C Density: 1.085 g/cm³ (20 °C) Flash Pt: 87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008357 CAS Number: 91-16-7 EINECS: 202-045-3 Merck Index: 12,10089 |