2,6-Dimethoxyphenol, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
D135550-100G D135550-25G
SIALD135550-100GEA 155 EUR
SIALD135550-100G SIALD135550-25G
2,6-Dimethoxyphenol, Sigma-Aldrich®
2,6-Dimethoxyphenol
Formula: C₈H₁₀O₃
Boiling Pt: 261 °C (1013 hPa)
Melting Pt: 52…56 °C
Storage Temperature: Ambient
MDL Number: MFCD00064434
CAS Number: 91-10-1
EINECS: 202-041-1