2,6-Dimethoxyphenol, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ300
W313718-100G-K W313718-1KG-K W313718-5KG-K
SIALW313718-100G-KEA 133 EUR
SIALW313718-100G-K SIALW313718-1KG-K SIALW313718-5KG-K
2,6-Dimethoxyphenol, Sigma-Aldrich®
2,6-Dimethoxyphenol
Formula: C₈H₁₀O₃
MW: 154.17 g/mol
Boiling Pt: 261
Melting Pt: 52…56 °C
Storage Temperature: Ambient
MDL Number: MFCD00064434
CAS Number: 91-10-1
EINECS: 202-041-1