3,4,5-Trimethoxyphenol, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Antiarol
M-Clarity™ quality level = MQ100
687162-25G 687162-5G
SIAL687162-25GEA 226 EUR
SIAL687162-25G SIAL687162-5G
3,4,5-Trimethoxyphenol, Sigma-Aldrich®
3,4,5-Trimethoxyphenol
Formula: C₉H₁₂O₄
Boiling Pt: 283 °C (1013 hPa)
Melting Pt: 147 °C
MDL Number: MFCD00008389
CAS Number: 642-71-7
EINECS: 211-387-2