Phthalic anhydride ≥99%
Supplier: Thermo Fisher Scientific
Danger
A14955.0B A14955.0I A14955.30
A14955.0BEA 37 EUR
A14955.0B A14955.0I A14955.30
Phthalic anhydride ≥99%
Phthalic anhydride
Purity: 99% CAS No.: 85-44-9 Molecular Formula: C8H4O3 Formula Weight: 148.12 Flash Point: 152°(305°F) Odour: Weak, characteristic Boiling Point: 295° subl. Beilstein Reference: 118515 Merck Reference: 14,7372 UN No.: 2214 MDL No.: MFCD00005918
Formula: C₈H₄O₃
MW: 148.12 g/mol
Boiling Pt: 295 °C (subl.)
Melting Pt: 130…134 °C
Density: 1.53
Flash Pt: 152 °C (305 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005918
CAS Number: 85-44-9
EINECS: 201-607-5
UN: 2214
ADR: 8 III
Merck Index: 13,07457