Phthalic anhydride, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
M-Clarity™ quality level = MQ200
125733-1KG
125733-2.5KG
SIAL125733-1KGEA
50.6
EUR
SIAL125733-1KG
SIAL125733-2.5KG
Phthalic anhydride, Sigma-Aldrich®
Phthalic anhydride
Formula:
C₈H₄O₃ MW: 148.12 g/mol Boiling Pt: 284 Melting Pt: 131 Density: 1.545 g/cm³ (20 °C) Flash Pt: 151 Storage Temperature: Ambient |
MDL Number:
MFCD00005918 CAS Number: 85-44-9 EINECS: 201-607-5 UN: 2214 ADR: 8 III Merck Index: 13,07457 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...