Phthalic anhydride, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ200

320064-25G 320064-2.5KG 320064-500G
SIAL320064-25GEA 37.8 EUR
SIAL320064-25G SIAL320064-2.5KG SIAL320064-500G
Phthalic anhydride, Sigma-Aldrich®
Phthalic anhydride
Formula: C₈H₄O₃
MW: 148.12 g/mol
Boiling Pt: 284
Melting Pt: 131
Density: 1.545 g/cm³ (20 °C)
Flash Pt: 151
Storage Temperature: Ambient
MDL Number: MFCD00005918
CAS Number: 85-44-9
EINECS: 201-607-5
UN: 2214
ADR: 8 III
Merck Index: 13,07457

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR