4-Phenoxyphenol
Supplier: Thermo Scientific
BTB14064.1
BTB14064.10
BTB14064.50
MAYBBTB14064.1EA
0
EUR
MAYBBTB14064.1
MAYBBTB14064.10
MAYBBTB14064.50
4-Phenoxyphenol
4-Phenoxyphenol
Formula:
C₁₂H₁₀O₂ Boiling Pt: 314 °C (1013 hPa) Melting Pt: 80…83 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002331 CAS Number: 831-82-3 EINECS: 212-611-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...