4-Phenoxyphenol, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

230669-25G 230669-5G
SIAL230669-25GEA 184 EUR
SIAL230669-25G SIAL230669-5G
4-Phenoxyphenol, Sigma-Aldrich®
4-Phenoxyphenol
Formula: C₁₂H₁₀O₂
Boiling Pt: 314 °C (1013 hPa)
Melting Pt: 80…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00002331
CAS Number: 831-82-3
EINECS: 212-611-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR