Pyrogallol, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
16040-1KG-R 16040-100G-R 16040-500G-R
SIAL16040-1KG-REA 730 EUR
SIAL16040-1KG-R SIAL16040-100G-R SIAL16040-500G-R
Pyrogallol, Sigma-Aldrich®
Pyrogallol
Formula: C₆H₆O₃
Boiling Pt: 309 °C (1013 hPa)
Melting Pt: 130…132 °C
Density: 1.453 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002192
CAS Number: 87-66-1
EINECS: 201-762-9
UN: 2811
ADR: 6.1,III
Merck Index: 13,08090
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=09c5271be2848ebfa6e3614bff56b329a59cb6fe&vid=db25c87d92a56d9ff5d36f04d6bbf59acf236659&prid=web&clid=SAPDAM