Benzenesulphonyl chloride, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Benzene sulphochloride
M-Clarity™ quality level = MQ100
108111-1KG
108111-25G
SIAL108111-1KGEA
87.1
EUR
SIAL108111-1KG
SIAL108111-25G
Benzenesulphonyl chloride, Sigma-Aldrich®
Benzenesulphonyl chloride
Formula:
C₆H₅ClO₂S Boiling Pt: 251 °C (1013 hPa) Melting Pt: 15 °C Density: 1.3842 g/cm³ (20 °C) Flash Pt: 119 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007426 CAS Number: 98-09-9 EINECS: 202-636-6 UN: 2225 ADR: 8,III Merck Index: 13,01072 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...