Benzene, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Cyclohexatriene
M-Clarity™ quality level = MQ200
401765-1L 401765-2L
SIAL401765-1LEA 138 EUR
SIAL401765-1L SIAL401765-2L
Benzene, Sigma-Aldrich®
Benzene
Formula: C₆H₆
MW: 78.11 g/mol
Boiling Pt: 80.1
Melting Pt: 5.5
Density: 0.878 g/cm³ (20 °C)
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00003009
CAS Number: 71-43-2
EINECS: 200-753-7
UN: 1114
ADR: 3 II
REACH: 01-2119447106-44
Merck Index: 13,01066
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d2c0146712ab33142aeeb3843b02381e6bda4028&vid=cae270bfe08ae6ab4b14d9e1775b89a82cfeec1f&prid=web&clid=SAPDAM