4-Vinylbenzoic acid, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
254738-1G 254738-5G
SIAL254738-1GEA 77.5 EUR
SIAL254738-1G SIAL254738-5G
4-Vinylbenzoic acid, Sigma-Aldrich®
4-Vinylbenzoic acid
Formula: C₉H₈O₂
Boiling Pt: 282 °C (1013 hPa)
Melting Pt: 115 °C
Density: 1.3 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00002569
CAS Number: 1075-49-6
EINECS: 214-053-4