Eugenol, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 4-Allyl-2-methoxyphenol, 5-Allyl-2-hydroxyanisole
M-Clarity™ quality level = MQ300
W246719-10KG-K W246719-1KG-K W246719-5KG-K
SIALW246719-10KG-KEA 1020 EUR
SIALW246719-10KG-K SIALW246719-1KG-K SIALW246719-5KG-K
Eugenol, Sigma-Aldrich®
Eugenol
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 255
Melting Pt: ∼–10 °C
Density: 1.0652 g/cm³ (20 °C)
Flash Pt: 110
Storage Temperature: Ambient
MDL Number: MFCD00008654
CAS Number: 97-53-0
EINECS: 202-589-1
Merck Index: 13,03931