4-Allylveratrole, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 4-Allylveratrole, Eugenol methyll ether, 4-Prop-2-enylveratrole, 4-Allyl-1,2-Dimethoxybenzene, 4-Allyl-1,2-dimethoxybenzene, Methyl eugenol, Eugenol methyl ether, Methyleugenol
M-Clarity™ quality level = MQ300
W247502-10KG-K W247502-1KG-K W247502-25KG-K
SIALW247502-10KG-KEA 741 EUR
SIALW247502-10KG-K SIALW247502-1KG-K SIALW247502-25KG-K
4-Allylveratrole, Sigma-Aldrich®
4-Allylveratrole
Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
Boiling Pt: 255
Melting Pt: –4 °C
Density: 1.036 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008652
CAS Number: 93-15-2
EINECS: 202-223-0