2-Allylphenol, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
A34805-100G A34805-500G
SIALA34805-100GEA 48.2 EUR
SIALA34805-100G SIALA34805-500G
2-Allylphenol, Sigma-Aldrich®
2-Allylphenol
Formula: C₉H₁₀O
Boiling Pt: 220 °C (1013 hPa)
Melting Pt: –2 °C
Density: 1.03 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002250
CAS Number: 1745-81-9
EINECS: 217-119-0
UN: 2922
ADR: 8,III