Phenethyl bromide, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: (2-Bromoethyl)benzene, 2-Phenylethyl bromide
M-Clarity™ quality level = MQ200
B65780-100G B65780-500G B65780-5G
SIALB65780-100GEA 46.8 EUR
SIALB65780-100G SIALB65780-500G SIALB65780-5G
Phenethyl bromide, Sigma-Aldrich®
Phenethyl bromide
Formula: BrCH₂CH₂C₆H₅
MW: 185.06 g/mol
Boiling Pt: 221
Melting Pt: –56 °C
Density: 1.37 g/cm³ (20 °C)
Flash Pt: 89
Storage Temperature: Refrigerator
MDL Number: MFCD00000240
CAS Number: 103-63-9
EINECS: 203-130-8