(2-Bromopropyl)benzene, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 2-Bromo-1-phenylpropane, (2-Bromopropyl)benzene
M-Clarity™ quality level = MQ100
B77401-25G B77401-5G
SIALB77401-25GEA 748 EUR
SIALB77401-25G SIALB77401-5G
(2-Bromopropyl)benzene, Sigma-Aldrich®
(2-Bromopropyl)benzene
Formula: C₆H₅CH₂CHBrCH₃
Boiling Pt: 107 °C (1013 hPa)
Density: 1.2908 g/cm³ (16 °C)
MDL Number: MFCD00000149
CAS Number: 2114-39-8
EINECS: 218-315-9