2,5-Dibromotoluene, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 1,4-Dibromo-2-methylbenzene
M-Clarity™ quality level = MQ100
250988-25G 250988-5G
SIAL250988-25GEA 57 EUR
SIAL250988-25G SIAL250988-5G
2,5-Dibromotoluene, Sigma-Aldrich®
2,5-Dibromotoluene
Formula: C₇H₆Br₂
MW: 249.93 g/mol
Boiling Pt: 236
Melting Pt: 6
Density: 1.8127 g/cm³ (19 °C)
MDL Number: MFCD00000090
CAS Number: 615-59-8
EINECS: 210-437-0