α,2-Dibromotoluene, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
2-Bromobenzyl bromide
M-Clarity™ quality level = MQ100
187070-100G
187070-25G
SIAL187070-100GEA
236
EUR
SIAL187070-100G
SIAL187070-25G
α,2-Dibromotoluene, Sigma-Aldrich®
α,2-Dibromotoluene
Formula:
C₇H₆Br₂ MW: 249.93 g/mol Boiling Pt: 253 Melting Pt: 34 Density: 1.881 g/cm³ (20 °C) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000173 CAS Number: 3433-80-5 EINECS: 222-334-8 UN: 2293 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...