2-Bromotoluene, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
B82006-100G B82006-25G
SIALB82006-100GEA 142 EUR
SIALB82006-100G SIALB82006-25G
2-Bromotoluene, Sigma-Aldrich®
2-Bromotoluene
Formula: C₇H₇Br
Boiling Pt: 181.5 °C (1013 hPa)
Melting Pt: –27 °C
Density: 1.4192 g/cm³ (20 °C)
Flash Pt: 79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000068
CAS Number: 95-46-5
EINECS: 202-421-7
UN: 3082
ADR: 9,III
Merck Index: 13,01424