α,4-Dibromotoluene, Sigma-Aldrich®
Supplier: Merck
Synonyms: 4-Bromobenzyl bromide, 1-Bromo-4-(bromomethyl)benzene
M-Clarity™ quality level = MQ100
112186-100G 112186-25G
SIAL112186-100GEA 65.2 EUR
SIAL112186-100G SIAL112186-25G
α,4-Dibromotoluene, Sigma-Aldrich®
α,4-Dibromotoluene
Formula: C₇H₆Br₂
Boiling Pt: 129…130 °C (18 mmHg)
Melting Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00000179
CAS Number: 589-15-1
EINECS: 209-636-5
UN: 2811
ADR: 6.1,II
Merck Index: 13,01393