4-Bromotoluene, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
B82200-100G B82200-500G B82200-5G
SIALB82200-100GEA 54.1 EUR
SIALB82200-100G SIALB82200-500G SIALB82200-5G
4-Bromotoluene, Sigma-Aldrich®
4-Bromotoluene
Formula: C₇H₇Br
Boiling Pt: 184 °C (1013 hPa)
Melting Pt: 24…27 °C
Density: 1.387 g/cm³ (20 °C)
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00000109
CAS Number: 106-38-7
EINECS: 203-391-8
UN: 3077
ADR: 9,III
Merck Index: 13,01424