α-Bromo-4-(trifluoromethoxy)toluene, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 1-(Bromomethyl)-4-(trifluoromethoxy)benzene, p-(Trifluoromethoxy)benzyl bromide, 4-(Trifluoromethoxy)benzyl bromide
M-Clarity™ quality level = MQ100
370622-1G 370622-5G
SIAL370622-1GEA 74.1 EUR
SIAL370622-1G SIAL370622-5G
α-Bromo-4-(trifluoromethoxy)toluene, Sigma-Aldrich®
α-Bromo-4-(trifluoromethoxy)toluene
Formula: C₈H₆BrF₃O
MW: 255.03 g/mol
Boiling Pt: 203
Melting Pt: 23…24 °C
Density: 1.58 g/cm³ (20 °C)
MDL Number: MFCD00061238
CAS Number: 50824-05-0
UN: 3261
ADR: 8 III