4-Isopropylbenzaldehyde, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Cuminaldehyde
M-Clarity™ quality level = MQ100
135178-100G 135178-5G
SIAL135178-100GEA 57.6 EUR
SIAL135178-100G SIAL135178-5G
4-Isopropylbenzaldehyde, Sigma-Aldrich®
4-Isopropylbenzaldehyde
Formula: C₁₀H₁₂O
MW: 148.2 g/mol
Boiling Pt: 235
Density: 0.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006953
CAS Number: 122-03-2
EINECS: 204-516-9
Merck Index: 13,02648