4-Propylbenzaldehyde, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ100

562882-1G 562882-5G
SIAL562882-1GEA 272 EUR
SIAL562882-1G SIAL562882-5G
4-Propylbenzaldehyde, Sigma-Aldrich®
4-Propylbenzaldehyde
Formula: C₁₀H₁₂O
Boiling Pt: 240 °C (1013 hPa)
Melting Pt: 19 °C
MDL Number: MFCD00041870
CAS Number: 28785-06-0
EINECS: 249-221-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR