3'-Chloropropiophenone ≥98%
Supplier: Thermo Fisher Scientific
Synonyms: m-Chloropropiophenone, 1-(3-Chlorophenyl)propan-1-one
A12542.14 A12542.22 A12542.36
A12542.14EA 39.5 EUR
A12542.14 A12542.22 A12542.36
3'-Chloropropiophenone ≥98%
3'-Chloropropiophenone
Purity: 98% CAS No.: 34841-35-5 Molecular Formula: C9H9ClO Formula Weight: 168.62 Flash Point: 110°(230°F) Melting Point: 45-47° Boiling Point: 123-124°/13mm MDL No.: MFCD00009925
Formula: C₉H₉ClO
MW: 168.62 g/mol
Boiling Pt: 123…124 °C (13 mmHg)
Melting Pt: 45…47 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009925
CAS Number: 34841-35-5
EINECS: 252-242-3