4-Chloro-3-(trifluoromethyl)aniline ≥99%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
5-Amino-2-chloro-α,α,α-trifluorotoluene, 1-Amino-4-chloro-3-(trifluoromethyl)benzene, 4-Chloro-α,α,α-trifluoro-m-toluidine, 5-Amino-2-chlorobenzotrifluoride
B21200.06
B21200.14
B21200.22
B21200.06EA
28.2
EUR
B21200.06
B21200.14
B21200.22
4-Chloro-3-(trifluoromethyl)aniline ≥99%
4-Chloro-3-(trifluoromethyl)aniline
Purity: 99%
CAS No.: 320-51-4
Molecular Formula: C7H5ClF3N
Formula Weight: 195.57
Flash Point: 134°(273°F)
Melting Point: 35-38°
Boiling Point: 85°/3mm
Beilstein Reference: 641588
UN No.: 2811
MDL No.: MFCD00007846
Formula:
C₇H₅ClF₃N MW: 195.57 g/mol Boiling Pt: 85 °C (3 mmHg) Melting Pt: 35…38 °C Flash Pt: 134 °C (273 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007846 CAS Number: 320-51-4 EINECS: 206-277-6 UN: 2811 ADR: 6.1 III |