6-Chloro-o-toluidine ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
L02629.06
L02629.14
L02629.06EA
79
EUR
L02629.06
L02629.14
6-Chloro-o-toluidine ≥98%
6-Chloro-o-toluidine
Purity: 98%
CAS No.: 87-63-8
Molecular Formula: C7H8ClN
Formula Weight: 141.60
Flash Point: 98°(208°F)
Refractive Index: 1.5775
Melting Point: 2°
Boiling Point: 215°
Beilstein Reference: 774624
UN No.: 3429
MDL No.: MFCD00007679
Formula:
C₇H₈ClN Boiling Pt: 215 °C Melting Pt: 2 °C Density: 1.174 Flash Pt: 98 °C (208 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007679 CAS Number: 87-63-8 EINECS: 201-759-2 UN: 3429 ADR: 6.1,III |