3-Methoxybenzyl chloride, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 1-(Chloromethyl)-3-methoxybenzene, 3-(Chloromethyl)anisole
209384-25G 209384-5G
SIAL209384-25GEA 48.9 EUR
SIAL209384-25G SIAL209384-5G
3-Methoxybenzyl chloride, Sigma-Aldrich®
3-Methoxybenzyl chloride
Formula: C₈H₉ClO
MW: 156.61 g/mol
Boiling Pt: 124
Density: 1.07 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000907
CAS Number: 824-98-6
EINECS: 212-541-1
UN: 1760
ADR: 8 II