3,5-Dichlorobenzyl alcohol

Supplier: Thermo Scientific

SPB00433.1 SPB00433EA
MAYBSPB00433.1EA 0 EUR
MAYBSPB00433.1 MAYBSPB00433EA
3,5-Dichlorobenzyl alcohol
3,5-Dichlorobenzyl alcohol
Formula: C₇H₆Cl₂O
Boiling Pt: 264 °C (1013 hPa)
Melting Pt: 82 °C
MDL Number: MFCD00004634
CAS Number: 60211-57-6
EINECS: 262-109-1
UN: 2250
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR