4-Chloro-m-cresol, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 4-Chloro-3-methylphenol, 2-Chloro-5-hydroxytoluene, 1-Chloro-4-hydroxy-2-methylbenzene, p-Chloro m-cresol
M-Clarity™ quality level = MQ100
C55402-5G C55402-100G C55402-500G
SIALC55402-5GEA 34.5 EUR
SIALC55402-5G SIALC55402-100G SIALC55402-500G
4-Chloro-m-cresol, Sigma-Aldrich®
4-Chloro-m-cresol
Formula: C₇H₇ClO
Boiling Pt: 235 °C (1013 hPa)
Melting Pt: 63…65 °C
Density: 0.9 g/cm³ (20 °C)
Flash Pt: 118 °C
Storage Temperature: Ambient
MDL Number: MFCD00002323
CAS Number: 59-50-7
EINECS: 200-431-6
UN: 3437
ADR: 6.1,II
Merck Index: 13,02152