3-Phenoxybenzyl chloride ≥97%
Supplier: Thermo Fisher Scientific
Danger
A15670.03 A15670.06 A15670.14
A15670.03EA 47.1 EUR
A15670.03 A15670.06 A15670.14
3-Phenoxybenzyl chloride ≥97%
3-Phenoxybenzyl chloride
Purity: 97% CAS No.: 53874-66-1 Molecular Formula: C13H11ClO Formula Weight: 218.69 Flash Point: 110°(230°F) Refractive Index: 1.5910 Boiling Point: 141-142°/5mm Beilstein Reference: 2212101 UN No.: 3265 MDL No.: MFCD00040866
Formula: C₁₃H₁₁ClO
MW: 218.68 g/mol
Boiling Pt: 141…142 °C (5 mmHg)
Density: 1.189
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040866
CAS Number: 53874-66-1
EINECS: 258-831-1
UN: 3265
ADR: 8 III